Packaging Buckets
Packaging Buckets
Actual product may vary
Fisherbrand
Packaging Buckets
Manufactured from polypropylene for strength and durability. Buerkle™ Packaging Buckets are ideal for transport, packaging and storing of a wide range of materials.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)TopHeight (Metric) With Closure
FKS118310911 L132 mm131 mm
Catalog number FKS11831091
Price (SGD)
8.00
PK
Add to cart
Capacity (Metric):
1 L
Diameter (Metric)Top:
132 mm
Height (Metric) With Closure:
131 mm
Price (SGD)
8.00
PK
Add to cart
Suitable for food or building material industries and non-aggressive chemicals in environmental protection, hosptials and household. Features strong handle and quality closure for product safety.
  • White polypropylene
  • Air-tight closure
  • Includes lid
Specifications
Product TypePackaging Bucket
Capacity (Metric)1 L
MaterialPolypropylene
ColorWhite
Diameter (Metric)Top132 mm
For Use With (Equipment)Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household
Height (Metric) With Closure131 mm
IncludesQuality Closure and Strong Lid
Quantity1 Ea.
Unit SizePK