Polypropylene Centrifuge Tubes
Polypropylene Centrifuge Tubes
Actual product may vary
Polypropylene Centrifuge Tubes
Polypropylene Centrifuge Tubes
Fisherbrand
Polypropylene Centrifuge Tubes
Printed with evenly spaced black graduations to enhance readability without losing precision. Fisherbrand™ Polypropylene Centrifuge Tubes provide high chemical resistance and RCF tolerance.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1182965050 mL
Catalog number FKS11829650
Price (SGD)
392.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
392.00
PK
Add to cart
  • Large white marking area makes it easy to label samples
  • Blue plug seal closure with a double start thread design provides a tight secure seal
  • Packed in sleeves, 25/sleeve
Specifications
Certifications/Compliance95 kPa
Closure ColorBlue
Capacity (Metric)50 mL
MaterialPolypropylene
For Use WithCentrifugation
Packaging25/Sleeve; Case of 500
SterilityNon-sterile
Closure TypePlug
Product TypeCentrifuge Tube
Unit SizePK