BioLite™ Microwell Plates
BioLite™ Microwell Plates
Actual product may vary
Fisher Catalogue
BioLite™ Microwell Plates
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Plates that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
Have Questions?
Catalog NumberNo. of WellsCulture AreaVolume (Metric) Well
FKS1182527569.6 cm23 mL
Catalog number FKS11825275
View Price:Sign in to see your account pricing. Need an account? Register with us today.
No. of Wells:
6
Culture Area:
9.6 cm2
Volume (Metric) Well:
3 mL
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated plates with lids suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Sterile per ISO 11137, SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• BSE/TSE Free
• RNase/DNase Free
• USP Class VI raw material
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidClear Closure Included
Culture Area9.6 cm2
Array2 x 3
ColorClear
Product TypeMulti Dish
Volume (Metric) Well3 mL
No. of Wells6
Volume (Metric) Working3 mL
SterilitySterile
Surface TreatmentCell Culture
Well ShapeFlat Bottom (F-well)
No. per Case50/Cs.
No. per Pack1 (individually wrapped)
Quantity50/Cs.
Shelf Life2 years
Unit SizePK