Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Actual product may vary
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Fisherbrand
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLidNo. per Case
FKS11816003SterileWith Lid160
FKS10580382Non-sterileWithout Lid180
FIS12-565-215SterileWithout Lid50
Catalog number FKS11816003
Price (SGD)
2,458.00
PK
Add to cart
Sterility:
Sterile
Lid:
With Lid
No. per Case:
160
Price (SGD)
2,458.00
PK
Add to cart
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
Features:
  • Conical bottom (V) microplates are ideal for sample recovery.
    • Applications include antibiotic screens, serological tests, storing and screening compounds, DNA libraries, and titration of solutions.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWith Lid
Array8 x 12
Packaging160 Cases
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
No. per Case160
No. per Pack10
Quantity160/Cs.
Unit SizePK