Series 1A Model 201NP Cooled Incubator, 201 L, Pre-coated Aluminum
Series 1A Model 201NP Cooled Incubator, 201 L, Pre-coated Aluminum
Actual product may vary
Fisher Catalogue
Series 1A Model 201NP Cooled Incubator, 201 L, Pre-coated Aluminum
Temperature Range: -10°C to +50°C
Have Questions?
Catalog NumberCapacity (Metric)
FKS11813420201 L
Catalog number FKS11813420
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
201 L
  • Range of cooled incubators with a white stove enamel steel exterior, foam insulation and a white pre-coated aluminum interior
  • Reliable and accurate temperature control
  • Standard and programmable models available
  • Programmable models offer multiple temperature control in multiple time zones
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload ±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls are easy to read and simple to use
  • Full range of optional extras available. including LED lighting, access porthole, inner perspex doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Certifications/ComplianceISO 9001:2008
Controller TypeProgrammable
DescriptionFreestanding incubator
Capacity (Metric)201 L
Chamber MaterialPre-coated Aluminum
Convection TechnologyFan assisted air circulation via premixing chamber
Dimensions (D x W) Shelf420 mm x 386 mm
LightingOptional LED
MaterialStove Enameled Mild Steel
No. of Shelves4
Temperature Range (Metric)-10°C to +50°C
TypeRefrigerated Incubator
DisplayDigital
InsulationFoamed polyurethane
Model201NP
RefrigerantCFC-free
Temperature Stability±0.1°C
Temperature Uniformity±0.5°C
Warranty2 Years
Width (English)20.5 in.
Width (English) Interior17.5 in.
Width (Metric)520 mm
Width (Metric) Interior445 mm
Height (English)56.3 in.
Height (English) Interior37.0 in.
Height (Metric)1430 mm
Height (Metric) Interior965 mm
Depth (English) Interior19.3 in.
Depth (Metric) Interior490 mm
Weight (Metric)65 kg
Electrical Requirements240 V, 50 Hz
Voltage240 V