Total Immersion Precision Thermometer
Total Immersion Precision Thermometer
Actual product may vary
Fisher Catalogue
Total Immersion Precision Thermometer
Use these thermometers for a variety of applications. Alla France™ Total Immersion Precision Thermometers are ideal for battery testing.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS11797429-20°C to +110°C
Catalog number FKS11797429
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-20°C to +110°C
  • Thermometers with yellow enamel backing
  • Red liquid filled
  • -20 to +110°C
  • Total immersion
  • 6 to 7mm diameter
  • Without certificate
Specifications
Bottle FillRed Liquid
For Use With (Application)General use
Immersion TypeTotal Immersion
Length (Metric)305 mm
MaterialGlass
Thermometer FillRed Liquid
Product TypeAlcohol Thermometer
Max. Temperature (Metric)110°C
Min. Temperature (Metric)-20°C
Temperature Range (Metric)-20°C to +110°C
Accuracy±1%
Unit SizeEA