Check Order Status
Foam Stand for Microtiter Tubes
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Foam Stand for Microtiter Tubes
Securely holds microtiter tubes during the mixing process. Foam Stands for Microtiter Tubes are for use with Vortex Mixers.
Have Questions?
Contact Us
Catalog Number
Color
FKS11792583
Black
Catalog number
FKS11792583
Price (SGD)
229.00
EA
Add to cart
Color:
Black
Price (SGD)
229.00
EA
Add to cart
Soft foam to prevent breakage
Plate not included
Specifications
Product Type
Foam Stand
Color
Black
For Use With (Equipment)
CLASSIC, ZX3, WIZARD, ZX4, TX4 Vortex Mixers
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link