Customizable Foam Stand for Vortex Mixer
Customizable Foam Stand for Vortex Mixer
Actual product may vary
Fisherbrand
Customizable Foam Stand for Vortex Mixer
Securely hold test tubes and vials in the configuration that works best for your needs. Customizable Foam Stands are compatible with Vortex Mixers.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1179257394 mm
Catalog number FKS11792573
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
94 mm
  • Soft foam to prevent breakage
  • Black in color
Specifications
Diameter (Metric)94 mm
For Use With (Equipment)CLASSIC, ZX3, WIZARD, ZX4, TX4
Unit SizeEA