Check Order Status
Class A USP Brown Graduated Measuring Pipettes
Actual product may vary
Fisherbrand
Class A USP Brown Graduated Measuring Pipettes
Provided with a USP dated batch identification certificate for uniformity.
Have Questions?
Change view
Catalog Number
Volume (Metric)
FKS11791516
10 mL
FKS11701526
25 mL
2 Options
Catalog number
FKS11791516
Price (SGD)
336.00
PK
Add to cart
Volume (Metric):
10 mL
Price (SGD)
336.00
PK
Add to cart
With silver diffusion ink for permanent markings
Brown graduation marks
Main point ring graduation
Graduated to tip
Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
Available in a range of volumes
Specifications
Certifications/Compliance
USP
Class
A
Material
AR-GLAS
For Use With (Application)
Volumetric accuracy in pipetting
Graduated
Yes
Graduation Color
Brown
Graduations
0.1 mL
Tolerance
0.03 mL
Volume (Metric)
10 mL
Length (Metric)
360 ±5 mm
Type
Measuring Pipet