Polypropylene Microtube Storage Box
Polypropylene Microtube Storage Box
Actual product may vary
Fisherbrand
Polypropylene Microtube Storage Box
Store 1.5 to 2.0 mL microtubes in secure boxes. Fisherbrand™ Polypropylene Microtube Storage Box features an easy-open lid and snap clasp, with molded grid lines and imprinted coordinates for easy identification. These boxes are natural colored and constructed of sturdy, autoclavable polypropylene.
Have Questions?
Catalog NumberColorPositionsArray
FKS11790334Natural10010 x 10
Catalog number FKS11790334
Price (SGD)
180.00
PK
Add to cart
Color:
Natural
Positions:
100
Array:
10 x 10
Price (SGD)
180.00
PK
Add to cart
Secure storage for microtube and cryogenic vials
  • Stores 1.5 to 2.0 mL microtubes or cryogenic vials
  • Each well has a well diameter of 12.3 mm
  • 50-well option ideal for sample or case study storage
  • 100-well option ideal for larger storage requirements
User-friendly construction
  • Easy-open lid with three-point hinges and a snap clasp for secure lid closure
  • Constructed of durable, autoclavable polypropylene
  • Molded grid lines on the lid and imprinted coordinates on the base and box
  • Available in natural hue or assorted colors for easy classification
  • Sturdy, flat boxes stack easily
Specifications
Product TypeMicrotube Rack
Closure IncludedYes
Array10 x 10
AutoclavableAutoclavable
ColorNatural
HoldsMicrotubes and Cryovials (1.5 to 2 mL)
MaterialPolypropylene
Positions100
StackableYes
For Use With (Application)Storage
For Use With (Equipment)Microcentrifuge Tube
No. per Pack5 Pack
Quantity5 Pack
Length (Metric)141 mm
Width (Metric)151 mm
Height (Metric)57 mm
Diameter (Metric) Well12.3 mm
Unit SizePK