Wire Gauze
Wire Gauze
Actual product may vary
Fisherbrand
Wire Gauze
Provides excellent support to beakers and other glassware during heating on ring stands.Fisherbrand™ Wire Gauze is a flat square manufactured from galvanized iron mesh.
Have Questions?
Catalog NumberType
FKS11789806Wire Gauze
Catalog number FKS11789806
Price (SGD)
107.00
PK
Add to cart
Type:
Wire Gauze
Price (SGD)
107.00
PK
Add to cart
  • Non-fraying edges
  • Durable and long-lasting
Specifications
TypeWire Gauze
Unit SizePK