Sterilin™ Dippas™
Sterilin™ Dippas™
Actual product may vary
Sterilin™ Dippas™
Sterilin™ Dippas™
Sterilin™ Dippas™
Fisherbrand
Sterilin™ Dippas™
Facilitate sample collection and transportation to the lab in a single container using Thermo Scientific™ Sterilin™ Dippas™ to reduce risk of cross-contamination.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Closure MaterialColor
FKS11788848100 mL383 mmMetalClear
Catalog number FKS11788848
Price (SGD)
1,591.00
PK
Add to cart
Capacity (Metric):
100 mL
Length (Metric):
383 mm
Closure Material:
Metal
Color:
Clear
Price (SGD)
1,591.00
PK
Add to cart
Facilitate sample collection and transportation to the lab in a single container using Thermo Scientific™ Sterilin™ Dippas™ to reduce risk of cross-contamination.
NOT AVAILABLE IN AMERICA
  • Individually wrapped and sterilized by gamma irradiation
  • Leak tested in accordance with BS EN ISO 6717
  • Available in either clear or blue polystyrene
  • Handle neatly snaps off after sample collection for ease of transportation
  • Choice of handle length to accommodate the majority of sampling applications
Specifications
Product TypeContainer
Closure IncludedYes
MaterialPolystyrene
SterilitySterile
Capacity (Metric)100 mL
Closure MaterialMetal
Closure TypeScrew Cap
ColorClear
DescriptionDippa, screw cap
Length (Metric)383 mm
Unit SizePK