Check Order Status
AR™ Glass Volumetric Pipettes
Actual product may vary
Fisherbrand
AR™ Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
Have Questions?
Change view
Catalog Number
Volume (Metric)
FKS11787971
10 mL
FKS11767971
2 mL
FKS11777971
5 mL
3 Options
Catalog number
FKS11787971
Price (SGD)
172.00
PK
Add to cart
Volume (Metric):
10 mL
Price (SGD)
172.00
PK
Add to cart
One mark
Amber stain graduation
Acc. to DIN EN ISO 648
Specifications
Class
B
Material
Glass
Graduated
Yes
Graduation Color
Brown
Quantity
6/Pk.
Tolerance
0.03 mL
Volume (Metric)
10 mL
Length (Metric)
450 mm
Type
Volumetric Pipet