DURAN™ Evaporating Dish, with spout
DURAN™ Evaporating Dish, with spout
Actual product may vary
DURAN™ Evaporating Dish, with spout
DURAN™ Evaporating Dish, with spout
Fisherbrand
DURAN™ Evaporating Dish, with spout
The wide, open shape of the evaporation dish creates a large liquid surface which favours the evaporation of excess solvents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11786223170 mL
FKS1177622390 mL
Catalog number FKS11786223
Price (SGD)
505.00
PK
Add to cart
Capacity (Metric):
170 mL
Price (SGD)
505.00
PK
Add to cart
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Spout for clean pouring
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
Specifications
AutoclavableAutoclavable
Capacity (Metric)170 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field
No. per Pack10/Pk.
Diameter (Metric)95 mm
Unit SizePK