3-Way Large Bore Stopcock
3-Way Large Bore Stopcock
Actual product may vary
Fisherbrand
3-Way Large Bore Stopcock
Perfect for handling higher flow volumes in process while using connections similar to luer systems. Cole-Parmer™ 3-Way Large Bore Stopcock is non-sterile and not approved or intended for medical, clinical, surgical or other patient oriented applications.
Have Questions?
Catalog NumberMaterial
FKS11785069Polycarbonate
Catalog number FKS11785069
Price (SGD)
241.00
PK
Add to cart
Material:
Polycarbonate
Price (SGD)
241.00
PK
Add to cart
  • Enhance fluid flow with a larger bore than standard luer stopcocks
  • Ideal for handling high flow volumes
Specifications
Description3-way flow
MaterialPolycarbonate
No. per Pack10 Pack
ConnectorsMale Luer Lock, Female Luer
Unit SizePK