Lid for Blood Collection Tray
Lid for Blood Collection Tray
Actual product may vary
Fisherbrand
Lid for Blood Collection Tray
Protect tray contents with a sturdy polystyrene cover. Fisherbrand™ Lid for Blood Collection Tray is custom designed for a snug fit and provides access to the handle in its vertical position.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS11780634420 x 240 x 205mm
Catalog number FKS11780634
Price (SGD)
280.00
EA
Add to cart
Dimensions (L x W x H):
420 x 240 x 205mm
Price (SGD)
280.00
EA
Add to cart
  • Can be secured with cable ties for extra security
  • Made of polystyrene and is stackable.
  • Measures 420 x 240 x 205mm
Specifications
Dimensions (L x W x H)420 x 240 x 205mm
For Use With (Equipment)Droplet™ Blood Collection Tray
Height (Metric)205mm
Length (Metric)420mm
Width (Metric)240mm