Search Thermo Fisher Scientific
Catalog Number | Length (English) | Thickness | Width (Metric) | Length (Metric) |
---|---|---|---|---|
FKS11779408 | 25 ft. | 24 μm | 46cm | 7.6 m |
FIS01-213-100 | 25 ft. | 18 μm | 30cm | 7.6 m |
FIS01-213-101 | 197 ft. | 18 μm | 30.5cm | 60 m |
FKS11769408 | 492 ft. | 18 μm | 30cm | 150 m |
FIS01-213-103 | 25 ft. | 24 μm | 46cm | 7.6 m |
FIS01-213-104 | 50 ft. | 24 μm | 46cm | 15.2 m |
FIS01-213-105 | 492 ft. | 24 μm | 46cm | 149.9 m |