3-Prong Clamps
3-Prong Clamps
Actual product may vary
Fisher Catalogue
3-Prong Clamps
Safely and securely holds articles in place in the laboratory. R&L Enterprises™ 3-Prong Clamps are manufactured from diecast aluminum alloy, actuated by nickel plated brass pummels and plastic headed nickel plated steel twin screw pivoting on a stainless steel pin.
Have Questions?
Catalog NumberJaw Type
FKS11775376Spring Loaded
Catalog number FKS11775376
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Jaw Type:
Spring Loaded
  • Jaws covered in chemically resistant plastic
  • For use with articles up to 90mm in diameter
  • Designed to hold articles within the jaws
  • Not recommended to hold articles by the points
  • Available with or without spring-loaded jaws
Specifications
Quantity1 Ea.
Type3-Prong Clamp
MaterialNickel, Plastic
Diameter (Metric)90 mm
Jaw TypeSpring Loaded
Unit SizeEA