1x1 image pixel for data collection
Acrylic 2-Compartment Dispensing Bin
Acrylic 2-Compartment Dispensing Bin
Actual product may vary
Fisherbrand
Acrylic 2-Compartment Dispensing Bin
Keep spare lab and office items easily accessible. Acrylic Dispensing Bins have large openings with hinged lids for easy filling and dispensing. The sturdy clear bins make it easy to monitor inventory. Two compartments help organize small parts.
Have Questions?
Catalog NumberColor
FKS11774834Transparent
Catalog number FKS11774834
Price (SGD)
364.00
EA
Add to cart
Color:
Transparent
Price (SGD)
364.00
EA
Add to cart
  • Ideal for pipette tips, tubes, fittings, syringe filters, septa and other small lab and office items
  • Made from 3 mm thick, transparent acrylic for at-a-glance inventory monitoring
  • Hinged lids over generous-sized openings for rapid filling and dispensing
  • Two compartments help to organize and categorize items
  • Flip-top covers keep products secure and dust free
  • Includes non-skid rubber feet
Specifications
TypeStorage Bin
MaterialAcrylic
ColorTransparent
ShapeRectangular
Quantity1 Ea.
Length (English)5.5 in.
Length (Metric)139 mm
Width (Metric)160 mm
Height (Metric)368 mm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link