Check Order Status
Grade 113 Cellulose Medium Filtering Qualitative Filter Paper
Actual product may vary
Fisherbrand
Grade 113 Cellulose Medium Filtering Qualitative Filter Paper
Filters fine particulates.
Have Questions?
Change view
Catalog Number
Format
Diameter (Metric)
DimensionsLxW
FKS11774263
Folded Disc
150 mm
FKS11734183
Disc
90 mm
FKS11714273
Folded Disc
320 mm
3 Options
Catalog number
FKS11774263
Price (SGD)
90.00
PK
Add to cart
Format:
Folded Disc
Diameter (Metric):
150 mm
Price (SGD)
90.00
PK
Add to cart
Pure grade
Made of cellulose fibers
Alpha content of nearly 100% according precise norms
Ash content:0.06%
Application
Used for liquids, fine particulates
Used in essential, industrial oil
Atmospheric analysis
Beverage industry
Food, chemical industry
Specifications
Description
Ash Content: 0.06%
Format
Folded Disc
Material
Cellulose
Particle Retention
5 to 8 μm
Pore Size
5 to 8 μm
For Use With (Application)
Suitable for fast filtration of liquids with fine particle
Grade
113
Quantity
100 Pk.
Thickness (Metric)
0.16 mm
Diameter (Metric)
150 mm