Gosselin™ Polypropylene 10mL Round-Base Tubes
Gosselin™ Polypropylene 10mL Round-Base Tubes
Actual product may vary
Fisherbrand
Gosselin™ Polypropylene 10mL Round-Base Tubes
Manufactured from polypropylene. Gosselin™ Polypropylene 10mL Round-Base Tubes stoppers are sold separately.
Have Questions?
Catalog NumberClosure Type
FKS11773294Wing Cap
Catalog number FKS11773294
Price (SGD)
740.00
PK
Add to cart
Closure Type:
Wing Cap
Price (SGD)
740.00
PK
Add to cart
  • Natural polypropylene 
  • Nominal capacity of 10 ML
  • Certified compliant with EC N°1935/2004, ISO 11137 : Nil
  • Without stopper/cap
Specifications
Certifications/ComplianceISO 11137
Closure ColorRed
Closure MaterialLDPE
ColorNatural
Capacity (Metric)10 mL
MaterialPolypropylene
For Use WithFor clinical, dairy, environment, industries, microbiology
Packaging250BG
Quantity1000 Carton
RimmedNo
SterilitySterile
Diameter (Metric)15.4 mm
Closure TypeWing Cap
Product TypeTest Tube
Unit SizePK