Search Thermo Fisher Scientific
| Catalog Number | Length (Metric) | Width (Metric) | Color |
|---|---|---|---|
| FKS11772644 | 38 m | 10 cm | Natural |
| FIS13-374-12 | 76 m | 10 cm | Natural |
| FIS13-374-16 | 76 m | 5 cm | Natural |
| FIS13-374-10 | 38 m | 10 cm | Natural |
Permeability
Effects of Common Reagents