SP Scienceware™ Pestle and Matched 1.5mL Tube
SP Scienceware™ Pestle and Matched 1.5mL Tube
Actual product may vary
Fisherbrand
SP Scienceware™ Pestle and Matched 1.5mL Tube
For resuspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials
Have Questions?
Catalog NumberSize
FKS11772324Pack of 100
Catalog number FKS11772324
Price (SGD)
589.00
PK
Add to cart
Size:
Pack of 100
Price (SGD)
589.00
PK
Add to cart
For resuspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials
  • Molded disposable polypropylene
  • Supplied with 1.5mL microcentrifuge tubes of same shape for optimal grinding
  • Pestle length: 3.5 in. (8.5cm)
Specifications
For Use With (Application)Resuspending bacterial cells, Precipitates of Nucleic Acids/Proteins and pellets of other material
AutoclavableAutoclavable
For Use With (Equipment)1.5mL Microcentrifuge tube
DisposableSingle-use
Length (English)3.34 in.
Capacity (Metric)8 mL
Length (Metric)85mm
MaterialPolypropylene
SizePack of 100
Unit SizePK