Stainless-steel Griffin-Style Beaker with Easy-Pour Rim
Stainless-steel Griffin-Style Beaker with Easy-Pour Rim
Actual product may vary
Fisherbrand
Stainless-steel Griffin-Style Beaker with Easy-Pour Rim
Assure accurate measuring with these beakers. Cole-Parmer™ Stainless-steel Griffin-Style Beaker with Easy-Pour Rim are great for higher temperatures and the general stirring or measuring of solutions.
Have Questions?
Catalog number FKS11772315
Price (SGD)
396.00
EA
Add to cart
Price (SGD)
396.00
EA
Add to cart
  • Durable, economical mixing capabilities
  • Medium gauge 304 SS
  • Measure volumes easily with internal graduation markers
Specifications
MaterialStainless Steel
Height (Metric)80 mm
Product TypeBeaker
Diameter (Metric) Outer66.6 mm
Unit SizeEA