UVGL-15 Compact UV Lamp
Product Image
Fisherbrand
UVGL-15 Compact UV Lamp
Use these lamps for variety of applications including fluorescence of stained gels and sterilization. UVP™ Compact UV Lamps are made of durable, lightweight plastic with an ergonomically designed handle.
Have Questions?
Catalog number FKS11768221
Price (SGD)
1,617.00
EA
Add to cart
Price (SGD)
1,617.00
EA
Add to cart
  • Lightweight, easy-to-handle lamp design
  • 4 watts
  • Use Compact UV Lamps with J-124 Lamp Stand for hands free operation
  • Use the lamps with C-10 Chromato-Vue Viewing Cabinet for viewing materials in a darkened environment
  • The Compact UV Lamp is available in a 12 Volt style (for cigarette lighter operation) which is designed for windshield curing (lamp part number 95-0019-14)
  • UV blocking eyewear recommended
Specifications
Voltage230 V
Wattage4 W
Wavelength365 nm
Case MaterialPlastic
Dimensions (L x W x H)198 x 71 x 53 mm
ModelUVGL-15 Compact UV Lamp
Product TypeUV Lamp
Unit SizeEA