DURAN™ Funnel, with long stem, Bunsen Funnel
DURAN™ Funnel, with long stem, Bunsen Funnel
Actual product may vary
DURAN™ Funnel, with long stem, Bunsen Funnel
DURAN™ Funnel, with long stem, Bunsen Funnel
Fisherbrand
DURAN™ Funnel, with long stem, Bunsen Funnel
A funnel enables a clean decanting and filtering of liquids. In addition, the Bunsen funnel has a long handle.
Have Questions?
Catalog NumberDiameter (Metric)Top
FKS1176617370 mm
Catalog number FKS11766173
Price (SGD)
1,063.00
PK
Add to cart
Diameter (Metric)Top:
70 mm
Price (SGD)
1,063.00
PK
Add to cart
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • With long stem
  • Bunsen funnel
  • Fired-on, highly durable white print
Specifications
Diameter (Metric)Top70 mm
MaterialBorosilicate Glass (3.3)
AutoclavableAutoclavable
For Use With (Application)Decanting and filtration of liquids of different densities; The spillage of substances is avoided; Filtering
Quantity10/Pk.
Product TypeNon-DIN/ISO size
Unit SizePK