1x1 image pixel for data collection
Conical Tube Holder
Conical Tube Holder
Actual product may vary
Conical Tube Holder
Conical Tube Holder
Conical Tube Holder
Fisherbrand
Conical Tube Holder
Allows user to clearly view the entire contents of tubes. Bel-Art™ Conical Tube Holder sets up in just seconds to hold either a 15 ml or a 50 ml conical tube.
Have Questions?
Catalog NumberPositions
FKS117652341
Catalog number FKS11765234
Price (SGD)
87.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Positions:
1
Price (SGD)
87.00
PK
Add to cart
Allows user to clearly view the entire contents of tubes. Bel-Art™ Conical Tube Holder sets up in just seconds to hold either a 15 ml or a 50 ml conical tube.
  • Versatile holder supports either a 15 or 50mL conical tube while allowing a view of the entire tube contents
  • Move tube and rack with one hand by pinching the uprights
  • Insert or remove tube from the holder with one hand
  • Steam autoclavable at 121°C (250°F)
  • Chemical-resistant polypropylene
Specifications
Product TypeTest Tube Single Rack
Array1 x 1
AutoclavableAutoclavable
ColorTransparent
HoldsTubes (15 or 50 mL)
MaterialPolypropylene
Positions1
ShapeCircular
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)15 and 50mL Conical Tubes
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link