Wheaton™ 47mm Filtration Assembly and Accessories
Wheaton™ 47mm Filtration Assembly and Accessories
Actual product may vary
Fisherbrand
Wheaton™ 47mm Filtration Assembly and Accessories
Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
FKS11764139Support Screen GasketPTFE
FKS11718859BaseGlass
Catalog number FKS11764139
Price (SGD)
124.00
PK
Add to cart
Type:
Support Screen Gasket
Material:
PTFE
Price (SGD)
124.00
PK
Add to cart
Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.
  • Includes 300mL graduated funnel
  • Includes PTFE support screen gasket
  • Includes anodized clamp and a No. 8 silicone stopper
  • Includes a 316 stainless steel support with 120 mesh screen
Specifications
For Use With (Equipment)Filtration Assembly (47 mm)
MaterialPTFE
No. per Pack6 Pk.
TypeSupport Screen Gasket
Diameter (Metric)47 mm
Unit SizePK