DeckWorks™ Low Binding Hinged Rack Barrier Tips
DeckWorks™ Low Binding Hinged Rack Barrier Tips
Actual product may vary
Fisherbrand
DeckWorks™ Low Binding Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures
Have Questions?
Catalog NumberVolume (Metric)
FKS117593250.1 to 10 μL
Catalog number FKS11759325
Price (SGD)
650.00
PK
Add to cart
Volume (Metric):
0.1 to 10 μL
Price (SGD)
650.00
PK
Add to cart
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
Specifications
Volume (Metric)0.1 to 10 μL
FilteredFiltered
SterilitySterile
ColorNatural
For Use With (Application)Used for quantitative assays in which sample binding can significantly alter results
FormatHinged Rack
Tip StyleLow Retention, Round
PackagingRack
GraduatedYes
Product LineDeckWorks
MaterialHDPE
Unit SizePK