1x1 image pixel for data collection
Grade 604 Surface Protection Paper
Grade 604 Surface Protection Paper
Actual product may vary
Fisherbrand
Grade 604 Surface Protection Paper
Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Packaging Type
FKS1175872250 m x 46 cmRoll
FKS11728742570 x 460 mmSheet
Catalog number FKS11758722
Price (SGD)
-
Dimensions (L x W):
50 m x 46 cm
Packaging Type:
Roll
  • Available in sheet or roll form
  • Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface
Specifications
Product TypeSurface Protection Paper
MaterialGrade 604 Paper
Length (Metric)50 m
Width (Metric)0.46 m
Dimensions (L x W)50 m x 46 cm
Backing MaterialPolyethylene
ColorWhite
Thickness0.25mm
Quantity1 roll
Packaging TypeRoll
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link