Gosselin™ HDPE Rectangular Sodium Thiosulfate Bottles
Gosselin™ HDPE Rectangular Sodium Thiosulfate Bottles
Actual product may vary
Fisherbrand
Gosselin™ HDPE Rectangular Sodium Thiosulfate Bottles
Sterilized by irradiation. Gosselin™ HDPE Rectangular Sodium Thiosulfate Bottles feature a screw cap.
Have Questions?
Catalog NumberColorShapeWidth (Metric)
FKS11758633NaturalSquare60mm
Catalog number FKS11758633
Price (SGD)
1,547.00
PK
Add to cart
Color:
Natural
Shape:
Square
Width (Metric):
60mm
Price (SGD)
1,547.00
PK
Add to cart
  • Pre-dosed with sodium thiosulfate which inhibits the action of chlorine, bromine and ozone dissolved in water
  • Sterilized by irradiation with peel away sterilisation certificate on the outer carton
  • 250mL to 500mL square HDPE or 500mL octagonal PETG - bulk packed
  • Bottles are screw cap assembled (tamper evidence not engaged)
Specifications
Capacity (Metric)250 mL
ColorNatural
Diameter (Metric) Inner Neck20 mm
MaterialHDPE
Length (Metric)60mm
SterilitySterile
Width (Metric)60mm
Closure ColorRed
Certifications/ComplianceEC N°1935/2004, ISO 11137
Closure IncludedYes
ShapeSquare
Height (Metric)155 mm
Closure MaterialPolyethylene
Closure TypeTamper-evident Screw Cap with Wad
Unit SizePK