Check Order Status
Gosselin™ Polystrene Petri Dish
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Gosselin™ Polystrene Petri Dish
Crystal polystyrene dishes, with optimal flatness.
Have Questions?
Contact Us
Catalog Number
Diameter (Metric)
Height (Metric)
Vent
FKS11758573
55 mm
14.2 mm
6
Catalog number
FKS11758573
Price (SGD)
772.00
PK
Add to cart
Diameter (Metric):
55 mm
Height (Metric):
14.2 mm
Vent:
6
Price (SGD)
772.00
PK
Add to cart
Appropriate for aseptic production
Sterilized by irradiation
Flat bottom
Specifications
Certifications/Compliance
EC N°1935/2004, ISO 11137, Aseptic
Color
Natural
Material
Polystyrene
Shape
Round
Sterility
Aseptic
Type
Petri Dish
Well Count
1
For Use With (Application)
For use in clinical, environment, industries, microbiology
Vent
6
Height (Metric)
14.2 mm
Diameter (Metric)
55 mm
Unit Size
PK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link