Class A Measuring Cylinders with NS Ground and Poly Stopper
Class A Measuring Cylinders with NS Ground and Poly Stopper
Actual product may vary
Fisherbrand
Class A Measuring Cylinders with NS Ground and Poly Stopper
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders with NS Ground and Poly Stopper are manufactured from durable DURAN™ glass tubing.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS1175667225 mL0.5 mL
Catalog number FKS11756672
Price (SGD)
206.00
PK
Add to cart
Capacity (Metric):
25 mL
Graduations:
0.5 mL
Price (SGD)
206.00
PK
Add to cart
  • Blue, main point graduation
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Base TypeHexagonal
Scale ColorBlue
ShapeRound
TypeGraduated Mixing Cylinder
Tolerance±0.25 mL
MaterialGlass
Certifications/ComplianceDIN EN ISO 4788
ClassA
Capacity (Metric)25 mL
Height (Metric)193 mm
IncludesPolyethylene Stopper
Stopper MaterialPolyethylene
Graduations0.5 mL
Unit SizePK