Metalware Range™ Stainless Steel Spoon One End Spatula
Metalware Range™ Stainless Steel Spoon One End Spatula
Actual product may vary
Fisherbrand
Metalware Range™ Stainless Steel Spoon One End Spatula
Nickel Electro™ Metalware Range™ Stainless Steel Spoon One End Spatula is a quick and easy to clean tool comprised of a spoon on one end and a spatula on the other end.
Have Questions?
Catalog NumberLength (Metric)
FKS11756617130 mm
Catalog number FKS11756617
Price (SGD)
156.00
PK
Add to cart
Length (Metric):
130 mm
Price (SGD)
156.00
PK
Add to cart
  • Quick to clean and has an attractive shiny appearance making it the material of choice for applications requiring sterile surfaces.
  • Immune to sterilizing solutions, most of the organic chemicals and dyestuffs, and a wide variety of inorganic chemicals
  • Non-magnetic
  • Bright finish
Specifications
Quantity5 Pk.
SterilitySterile
AutoclavableAutoclavable
Capacity (Metric)2 mL
Product TypeChattaway Double Spatula
FinishBright
For Use With (Application)Sterile Applications
Width (Metric)10 mm
Dimensions (L x W)130 x 10 mm
Temperature (Metric) Max-196°C to +550°C
MaterialNickel
Length (Metric)130 mm
Unit SizePK