SA Poxygrid Centrifuge Tube Rack
SA Poxygrid Centrifuge Tube Rack
Actual product may vary
Fisherbrand
SA Poxygrid Centrifuge Tube Rack
Holds tubes that cannot stand upright by themselves. Rack allows for convenient transport and storage of up to four 500 mL conical-bottom centrifuge tubes.
Have Questions?
Catalog NumberHolds
FKS11755234Tubes (500 mL)
Catalog number FKS11755234
Price (SGD)
207.00
PK
Add to cart
Holds:
Tubes (500 mL)
Price (SGD)
207.00
PK
Add to cart
Holds tubes that cannot stand upright by themselves. Rack allows for convenient transport and storage of up to four 500 mL conical-bottom centrifuge tubes.
  • Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis
  • Temperature range: -223ºC (-369ºF) to 121ºC (250ºF)
  • Autoclavable
Specifications
Product TypeCentrifuge Tube Rack
Array1 x 4
AutoclavableAutoclavable
ColorBlue
HoldsTubes (500 mL)
MaterialEpoxy-coated Steel
Positions4
ShapeSquare
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)Conical centrifuge tubes
Dimensions (L x W x H)8 x 8 x 3 in. (20.3 x 20.3 x 7.6 cm)
Length (Metric)20.3 cm
Length (English)8 in.
Width (Metric)20.3 cm
Width (English)8 in.
Height (Metric)7.6 cm
Height (English)3 in.
Unit SizePK