Polypropylene Mixer Shaft Assembly
Polypropylene Mixer Shaft Assembly
Actual product may vary
Fisherbrand
Polypropylene Mixer Shaft Assembly
Use these plastic paddle assemblies for mixing most acids and corrosive solutions. Polypropylene Mixer Shaft Assemblies provide better chemical resistance than stainless steel alone.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1174998870 mm
Catalog number FKS11749988
Price (SGD)
339.00
EA
Add to cart
Diameter (Metric):
70 mm
Price (SGD)
339.00
EA
Add to cart
Choose from three plastics—polypropylene, polyethylene, or PTFE—according to the chemical compatibility of your solutions. For mixing of highly viscous liquids.
Specifications
Diameter (Metric)70 mm
Diameter (Metric) Shaft8 mm
Length (Metric) Handle350 mmL
Quantity1 Pack
TypeMixer Shaft Assembly
Unit SizeEA