Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Actual product may vary
Fisher Catalog
Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Maintain integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials: Polypropylene Caps are Chemically inert, suitable for strong solvents and high-purity chemicals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Liner)Closure Size
FKS11748819Metal Foil12.3 mm
FIS03-341-25DFoamed Polyethylene12.3 mm
FIS03-341-25LMetal Foil13.1 mm
FIS03-341-25KFoamed Polyethylene13.1 mm
FIS03-341-25EMetal Foil12.3 mm
Catalog number FKS11748819
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material (Liner):
Metal Foil
Closure Size:
12.3 mm
Maintain integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials: Polypropylene Caps are Chemically inert, suitable for strong solvents and high-purity chemicals.
  • Chemically inert, suitable for strong solvents and high-purity chemicals
  • Variety of cap liners available to maintain integrity of seal
  • Background counts are consistent and low; ultraviolet transmission is high
  • With screw caps attached to vials
  • Vials are made from clear glass; caps are white plastic.
Specifications
Closure ColorWhite
Material (Liner)Metal Foil
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialBorosilicate Glass
Volume (Metric)20 mL
For Use With (Application)Good barrier properties and resistance to hydrocarbons, oils, ketones and alcohols
Height (English)2.4 in.
Packaging TypeCellular Trays
Volume (English)0.67 oz.
Volume (Metric) Total20 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap
Height (Metric)61 mm
Certifications/ComplianceASTM E438, Type I, Class A
TypeScintillation Vial
Closure Size12.3 mm
Unit SizePK