Carbon Steel Scalpel Blades
Carbon Steel Scalpel Blades
Actual product may vary
Fisherbrand
Carbon Steel Scalpel Blades
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeLength (Metric)
FKS11748373# 1120 mm
FKS11702734# 2330 mm
Catalog number FKS11748373
Price (SGD)
63.00
PK
Add to cart
Size:
# 11
Length (Metric):
20 mm
Price (SGD)
63.00
PK
Add to cart
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Blade MaterialCarbon Steel
Product TypeScalpel Blade
Size# 11
Length (Metric)20 mm
For Use With (Equipment)The # 11 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
DisposableSingle-use
Certifications/ComplianceBS EN 7740
Quantity10/Pk.
Unit SizePK