316 Stainless Steel Turbine Flowmeter
316 Stainless Steel Turbine Flowmeter
Actual product may vary
Fisherbrand
316 Stainless Steel Turbine Flowmeter
Have Questions?
Catalog NumberFlow Rate Range
FKS117472388 to 76 LPM
Catalog number FKS11747238
Price (SGD)
6,695.00
EA
Add to cart
Flow Rate Range:
8 to 76 LPM
Price (SGD)
6,695.00
EA
Add to cart
Specifications
TypeTurbine Flowmeter
Accuracy±2% (Reading)
Float MaterialStainless Steel (316)
Pressure Rating1500 psig
Connections1/8 in. NPT (F)
Flow Rate Range8 to 76 LPM
For Use With (Application)Excellent selection for most basic industrial flow metering applications
Height (Metric)50 mm
Power Supply DetailsTwo 3 V Lithium Batteries
Repeatability±0.1%
Width (Metric)110 mm
Temperature (Metric)60°C
Weight (Metric)3.2 kg
Unit SizeEA