GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets
GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets
Actual product may vary
Fisherbrand
GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets
Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
FKS11747194SachetEZ Anaerobe Container System
Catalog number FKS11747194
Price (SGD)
754.00
Pack
Add to cart
Type:
Sachet
For Use With (Equipment):
EZ Anaerobe Container System
Price (SGD)
754.00
Pack
Add to cart
  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Specifications
TypeSachet
FormatSachet
For Use With (Application)Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment)EZ Anaerobe Container System
Product LineGasPak
Quantity20 Sachets/Box
Unit SizePack
Contents & Storage
2°C to 25°C