Mixed Cellulose Ester Membranes Sterile Filter
Mixed Cellulose Ester Membranes Sterile Filter
Actual product may vary
Fisherbrand
Mixed Cellulose Ester Membranes Sterile Filter
Fisherbrand™ Mixed Celluloses Esters Membranes Sterile Filters are composed of cellulose acetate and cellulose nitrate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeColor
FKS117452850.45 μmBlack
FKS117352850.45 μmWhite
Catalog number FKS11745285
Price (SGD)
291.00
PK
Add to cart
Pore Size:
0.45 μm
Color:
Black
Price (SGD)
291.00
PK
Add to cart
  • Widely used membranes in analytical and research applications
  • Naturally hydrophilic, biologically inert and offer thermal stability
  • Characterized by smoother and more uniform surface than pure nitrocellulose membrane which facilitates particles detection, and high retention level
  • Gridded membranes are used in many microbiological techniques include colony counting after incubation
Specifications
Bubble Point2.23 bar
ColorBlack
Filter TypeMicrofilter
MaterialMCE
Pore Size0.45 μm
SterilitySterile
Flow Rate60 mL/min./cm2
For Use With (Application)Microdialysis of DNA and Proteins, clarification of aqueous solutions during microbiological analysis, identification of Nucleic Acids
Gas Flow Rate5 L/min.
Grid ColorOff-white
HydrophobicityHydrophilic
Quantity100/Pk.
TypeGridded Disc
WettabilityYes
Diameter (Metric)47 mm
Unit SizePK