Straight Containers with Hinged Cap
Straight Containers with Hinged Cap
Actual product may vary
Fisherbrand
Straight Containers with Hinged Cap
Provides safe and secure storage, with optimized volume for milk sampling. Gosselin™ Straight Containers with Hinged Cap are manufactured from either clear or blue polystyrene.
Have Questions?
Catalog NumberCapacity (Metric)Closure ColorNo. per Case
FKS1174315490 mLNatural350/Cs.
Catalog number FKS11743154
Price (SGD)
369.00
Pack
Add to cart
Capacity (Metric):
90 mL
Closure Color:
Natural
No. per Case:
350/Cs.
Price (SGD)
369.00
Pack
Add to cart
  • Shock resistant
  • Auto-stable hinged cap
  • With or without writing area
Specifications
Product TypeContainer
Closure IncludedYes
GraduatedYes
MaterialPolypropylene
ShapeStraight, Cylindrical
SterilityNon-sterile
Capacity (Metric)90 mL
Certifications/ComplianceEC N°1935/2004
Closure ColorNatural
Closure TypeHinged Cap
ColorNatural
Diameter (Metric)43 mm
Graduations80 mL
Height (Metric)80 mm
IncludesNatural Polypropylene hinged cap
No. per Case350/Cs.
Unit SizePack