Orion™ Single Junction Combination pH Electrodes
Orion™ Single Junction Combination pH Electrodes
Actual product may vary
Fisherbrand
Orion™ Single Junction Combination pH Electrodes
Confidently measure your samples with the Orion Silver Chloride pH electrodes.
Have Questions?
Catalog NumberConnector TypesJunctionTypeMaterial
FKS11742837BNCCeramicCombination pH ElectrodeGlass Body
Catalog number FKS11742837
Price (SGD)
582.00
EA
Add to cart
Connector Types:
BNC
Junction:
Ceramic
Type:
Combination pH Electrode
Material:
Glass Body
Price (SGD)
582.00
EA
Add to cart
Thermo Scientific™ Orion™ Silver Chloride pH Electrodes are available in different body materials with either refillable or low maintenance gel options too. The 9142BN is a refillable electrode with chemically resistant glass body best suited for general purpose samples, including solvents, while the 9146BN and 9147BN contain a low-maintenance gel that is best suited for field or plant use, and also educational institutes, where minimal maintenance is preferred. The 9147BN also comes with a built-in ATC.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required 
Electrode fill solution for 9142BN
900011     4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 9142BN
900011     4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
6 months from date of purchase
Specifications
DescriptionRefillable combination pH electrode with glass body
Connector TypesBNC
JunctionCeramic
MaterialGlass Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypeCombination pH Electrode
For Use With (Equipment)Russell meters
Storage RequirementsStore in Orion pH electrode storage solution 910001
Temperature (English) Operating32°F to 176°F
RefillableRefillable
Unit SizeEA