Check Order Status
M3 Pleat-Style Face Masks
Actual product may vary
Fisherbrand
M3 Pleat-Style Face Masks
Pleat-style mask available with earloops or soft ties. Suggested for use in ISO Class 3 and higher cleanroom environments.
Have Questions?
Change view
Catalog Number
Fastening Type
Width (Metric)
Width (English)
FKS11742473
Earloops
18 cm
7.1 in.
FIS18-999-4774
Earloops
18 cm
7.1 in.
FIS18-999-4775
Ties
18 cm
7.1 in.
3 Options
Catalog number
FKS11742473
Price (SGD)
-
Fastening Type:
Earloops
Width (Metric):
18 cm
Width (English):
7.1 in.
Inner facing made of white BiCoSof™
Nose piece made from fully enclosed, soft, malleable aluminum
Outer facing made from clear polyethylene apertured film
Specifications
Product Type
Cleanroom Face Mask
Color
White
Fastening Type
Earloops
Material
Polyethylene
Size
Standard
Sterility
Non-sterile
Style
Pleated
Cleanroom Class
Class 1 (ISO 3)
Disposable
Single-use
Filter Efficiency
99.5% (PFE), 98.7% (BFE)
For Use With (Application)
Semiconductor, Disk Drive, Data Storage, Integrated Circuits
Latex Free
Yes
Model
62465
Pore Size
0.1 μm (PFE), 3 μm (BFE)
Series
Kimtech™ M3
Width (English)
7.1 in.
Width (Metric)
18 cm