Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FKS11742182 | 500 mL |
| FKS11722182 | 250 mL |
Thanks to their exceptional properties, such as excellent chemical resistance, nearly inert behaviour and high usage temperature, the original DURAN™ Erlenmeyer flasks are ideal for use in laboratories. Moreover, the uniform wall thickness, minimal thermal expansion and the resultant high thermal shock resistance are outstanding characteristics. Therefore, DURAN™ Erlenmeyer flasks are particularly suited to use with very high temperatures and fast temperature changes.