1x1 image pixel for data collection
DURAN™ Weighing Bottle, with ground lid, high form
DURAN™ Weighing Bottle, with ground lid, high form
Actual product may vary
Fisherbrand
DURAN™ Weighing Bottle, with ground lid, high form
Weighing glasses are suitable for weighing substances. The matching lid prevents unintentional distribution of the weighing product.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1174096445 mL
Catalog number FKS11740964
Price (SGD)
1,383.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
45 mL
Price (SGD)
1,383.00
PK
Add to cart
In order to carry out reactions in a controlled manner, it is essential to know the quantities of substances required and to weigh the corresponding substances accurately. A weighing glass can be used for weighing. The associated lid allows the weighing pan to be closed and thus prevents, for example, turbulence of the weighed-in solids during transport. Since even the smallest contamination of the weighing glass can have an effect on the weighing inheritance, the glass in the area of the ground joint and the lid knob are so pronounced that they can be grasped and moved with forceps or tweezers.
  • Close-fitting lid prevents any sample loss during transport after weighing
  • Fired-on, highly durable white ceramic
Specifications
Capacity (Metric)45 mL
DescriptionHigh form
For Use With (Application)Weighing; Storage and transport; Available in low and high forms
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
AutoclavableAutoclavable
Diameter (Metric)38 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link