Class AS Graduated Pipettes
Class AS Graduated Pipettes
Actual product may vary
Class AS Graduated Pipettes
Class AS Graduated Pipettes
Fisherbrand
Class AS Graduated Pipettes
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes are available in either blue or amber graduation.
Have Questions?
Catalog NumberVolume (Metric)GraduationsGraduation ColorTolerance
FKS117408141 mL0.1 mLBlue0.007 mL
Catalog number FKS11740814
Price (SGD)
227.00
PK
Add to cart
Volume (Metric):
1 mL
Graduations:
0.1 mL
Graduation Color:
Blue
Tolerance:
0.007 mL
Price (SGD)
227.00
PK
Add to cart
  • Acc. to DIN EN ISO 835
  • Main point ring graduation (in either blue or amber)
  • Graduated to tip, zero on tip, type 3
  • With cotton plug end as standard
  • Conformity certified, with dated batch identification
Specifications
ClassAS
MaterialAR-GLAS
GraduatedYes
Graduation ColorBlue
Graduations0.1 mL
Quantity12 Pack
Tolerance0.007 mL
Volume (Metric)1 mL
Length (Metric)360 ±5 mm
TypeGraduated Pipet
Unit SizePK