Check Order Status
Cellulose and Polyester Filter Cartridge
Actual product may vary
Fisherbrand
Cellulose and Polyester Filter Cartridge
Have Questions?
Catalog number
FKS11740649
Price (SGD)
99.00
EA
Add to cart
Price (SGD)
99.00
EA
Add to cart
Specifications
Flow Rate
15 GPM
Material
Cellulose, Polyester
Quantity
1 Pack
Type
Filter Cartridge
Length (Metric)
254 mm