Floor Stand Wiper Dispenser
Floor Stand Wiper Dispenser
Actual product may vary
Floor Stand Wiper Dispenser
Use this mobile dispenser use to quickly and easily dispense paper wipers from a large roll. Floor Stand Wiper Dispenser is a large, sturdy, blue tubular steel dispenser that can be moved around as a free-standing unit. Dispenser is ideal for busy food areas and helping to maintain hygiene standards.
Have Questions?
Catalog NumberMounting
FKS11738188Freestanding
Catalog number FKS11738188
Price (SGD)
176.00
EA
Add to cart
Mounting:
Freestanding
Price (SGD)
176.00
EA
Add to cart
  • One blue Kimberly-Clark Professional™ floor standing large roll wiper dispenser
  • A mobile, free-standing blue roll dispenser that can be moved around the workplace
  • Blue, tubular steel Kimberly-Clark dispenser for large wiper rolls in busy environments
  • Our portable dispenser is ideal for use in busy food areas, where the strictest hygiene standards are required
  • Compatible with a variety of Kimberly-Clark Professional™, WypAll™ and Kimtech™ blue roll, wipers and hand towels
Specifications
Product TypeFloor Stand Wiper Dispenser
ColorBlue
Dimensions (L x W x H)96 x 51.5 x 55 cm
Length (English)37.8 in.
Length (Metric)96 cm
Width (English)20.3 in.
Width (Metric)51.5 cm
Height (English)21.7 in.
Height (Metric)55 cm
HoldsWipes
MaterialSteel
MountingFreestanding
No. of Compartments1
Product StyleSide Loading
For Use With (Application)To maintain hygiene
IncludesTubular Steel Dispenser
Quantity1 Ea.
Unit SizeEA