GasPak™ EZ Gas Generating Systems Incubation Containers
GasPak™ EZ Gas Generating Systems Incubation Containers
Actual product may vary
Fisherbrand
GasPak™ EZ Gas Generating Systems Incubation Containers
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Have Questions?
Catalog NumberHoldsSize
FKS11737194Petri Dishes (30 to 33)Large
Catalog number FKS11737194
Price (SGD)
1,401.00
EA
Add to cart
Holds:
Petri Dishes (30 to 33)
Size:
Large
Price (SGD)
1,401.00
EA
Add to cart
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
  • Non-breakable, chemical resistant containers
  • Removable Petri dish racks are available (sold separately)
Specifications
TypeEZ Incubation Container
Closure TypeEasy close latches
DisposableReusable
For Use With (Application)Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
HoldsPetri Dishes (30 to 33)
Product LineGasPak
SizeLarge
Unit SizeEA
Contents & Storage
2°C to 25°C